Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Igha
- Entrez Gene ID
-
314487
- Gene Name
- immunoglobulin heavy chain, alpha
- Gene Aliases
-
-
- Chromosome Location
-
Chr.6: 138054656 - 138056613 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC087137.1 |
- |
- |
322 |
137 |
- Gene Symbol
- LOC500737
- Entrez Gene ID
-
500737
- Gene Name
- immunoglobulin heavy chain variable region
- Gene Aliases
-
-
- Chromosome Location
-
Chr.6: 140101804 - 140102401 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
CO559097.1 |
- |
- |
316 |
137 |
View Full Product Details