Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bin2a
Entrez Gene ID
494244
Gene Name
beta-galactosidase-like protein
Gene Aliases
Glb1l4
Chromosome Location
Chr.8: 27935385 - 27974006 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009524.1 NP_001009524.1 2 - 3 358 160
XM_006242737.2 XP_006242799.1 2 - 3 236 160
GenBank mRNA AF299297.1 - 2 - 3 358 160
AF299298.1 - 2 - 3 212 160
AF414431.1 - 2 - 3 212 160
AY040870.1 - 2 - 3 358 160

View Full Product Details