Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ka11
Entrez Gene ID
450226
Gene Name
type I keratin KA11
Gene Aliases
-
Chromosome Location
Chr.10: 88141667 - 88144624 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008750.1 NP_001008750.1 3 - 4 662 132

View Full Product Details