Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RT1-S2
Entrez Gene ID
24994
Gene Name
RT1 class Ib, locus S2
Gene Aliases
RT1.S1
Chromosome Location
Chr.20: 3153057 - 3165495 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008857.1 NP_001008857.1 4 - 5 492 137
XM_008772705.1 XP_008770927.1 4 - 5 3087 137

View Full Product Details