Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rnase11
Entrez Gene ID
497651
Gene Name
ribonuclease, RNase A family, 11 (non-active)
Gene Aliases
Raj1
Chromosome Location
Chr.15: 27973371 - 27977885 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012476.2 NP_001012494.1 2 - 2 594 72
XM_006251899.2 XP_006251961.1 2 - 2 830 72
XM_008770562.1 XP_008768784.1 2 - 2 602 72
GenBank mRNA BC128714.1 - 2 - 2 594 72

View Full Product Details