The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014023.1 | NP_001014045.1 | 5 - 6 | 881 | 68 |
XM_006229912.2 | XP_006229974.1 | 5 - 6 | 1367 | 68 | |
GenBank mRNA | BC078926.1 | - | 5 - 6 | 881 | 68 |