Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Hrk
- Entrez Gene ID
-
117271
- Gene Name
- harakiri, BCL2 interacting protein
- Gene Aliases
-
Bid3, Dp5
- Chromosome Location
-
Chr.12: 44008877 - 44029211 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
D83697.1 |
- |
2 - 2 |
4952 |
89 |
View Full Product Details