The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173329.2 | NP_775451.2 | 2 - 3 | 250 | 82 |
XM_006253367.1 | XP_006253429.1 | 2 - 3 | 294 | 82 | |
GenBank mRNA | U16685.1 | - | 2 - 3 | 229 | 82 |
U16686.1 | - | 2 - 3 | 250 | 82 |