Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
Vom2r58
Entrez Gene ID
501793
Gene Name
vomeronasal 2 receptor, 58
Gene Aliases
-
Chromosome Location
Chr.12: 5057134 - 5201595 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099479.1 NP_001092949.1 5 - 5 1542 155

Gene Symbol
Vom2r59
Entrez Gene ID
288396
Gene Name
vomeronasal 2 receptor, 59
Gene Aliases
RGD1566381
Chromosome Location
Chr.12: 3969493 - 4032641 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099466.1 NP_001092936.1 5 - 5 1542 155

View Full Product Details