Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aadacl3
Entrez Gene ID
313686
Gene Name
arylacetamide deacetylase-like 3
Gene Aliases
RGD1563257
Chromosome Location
Chr.5: 162323373 - 162331598 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191768.1 NP_001178697.1 1 - 2 170 74

View Full Product Details