The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001191963.1 | NP_001178892.1 | 4 - 5 | 3370 | 73 |
XM_008761470.1 | XP_008759692.1 | 5 - 6 | 3377 | 73 |