The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271180.1 | NP_001258109.1 | 5 - 6 | 827 | 72 |
GenBank mRNA | FQ213935.1 | - | 5 - 6 | 818 | 72 |