Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GRS1
Entrez Gene ID
852418
Gene Name
Gene Aliases
YBR121C
Chromosome Location
Chr.II: 481364 - 483367 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178469.2 NP_009679.2 1 - 1 1946 96

View Full Product Details