Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ERP2
Entrez Gene ID
851226
Gene Name
Gene Aliases
YAL007C
Chromosome Location
Chr.I: 137698 - 138345 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178152.1 NP_009395.1 1 - 1 542 93

View Full Product Details