Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RIP1
Entrez Gene ID
856689
Gene Name
Gene Aliases
YEL024W
Chromosome Location
Chr.V: 107260 - 107907 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178839.3 NP_010890.3 1 - 1 545 77

View Full Product Details