Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GCN20
Entrez Gene ID
850561
Gene Name
Gene Aliases
YFR009W
Chromosome Location
Chr.VI: 162488 - 164746 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179974.1 NP_116664.1 1 - 1 2150 95

View Full Product Details