Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ERV14
Entrez Gene ID
852826
Gene Name
Gene Aliases
YGL054C
Chromosome Location
Chr.VII: 400871 - 401287 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180919.1 NP_011461.1 1 - 1 347 88

View Full Product Details