Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALO1
Entrez Gene ID
854888
Gene Name
Gene Aliases
YML086C
Chromosome Location
Chr.XIII: 95791 - 97371 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182445.1 NP_013624.1 1 - 1 1350 66

View Full Product Details