Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
INN1
Entrez Gene ID
855570
Gene Name
Gene Aliases
YNL152W
Chromosome Location
Chr.XIV: 346313 - 347542 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182990.1 NP_014247.1 1 - 1 1154 80

View Full Product Details