Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OST2
Entrez Gene ID
854270
Gene Name
Gene Aliases
YOR103C
Chromosome Location
Chr.XV: 516449 - 516841 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183522.1 NP_014746.2 1 - 1 253 67

View Full Product Details