Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
PFY1
Entrez Gene ID
854289
Gene Name
Gene Aliases
CLS5, PRF1, YOR122C
Chromosome Location
Chr.XV: 552298 - 552887 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183541.3 NP_014765.3 1 - 1 48 79

View Full Product Details