Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COT1
Entrez Gene ID
854494
Gene Name
Gene Aliases
YOR316C
Chromosome Location
Chr.XV: 906236 - 907555 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183736.3 NP_014961.3 1 - 1 1251 72

View Full Product Details