Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YTA6
Entrez Gene ID
856031
Gene Name
Gene Aliases
YPL074W
Chromosome Location
Chr.XVI: 415763 - 418027 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183888.1 NP_015251.1 1 - 1 2155 106

View Full Product Details