Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ITM2B
Entrez Gene ID
595120
Gene Name
integral membrane protein 2B
Gene Aliases
Bri, imBRI2
Chromosome Location
Chr.11: 19349088 - 19377931 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042375.1 NP_001035834.1 6 - 6 754 74
XM_005652339.3 XP_005652396.2 - 1001 74
GenBank mRNA AK232104.1 - 6 - 6 917 74
AK233398.1 - 6 - 6 933 74
AK347736.1 - 6 - 6 921 74
AK389243.1 - 6 - 6 939 74
AK391720.1 - 6 - 6 910 74
AK392962.1 - 6 - 6 926 74
AK395823.1 - 6 - 6 903 74
AK397137.1 - 6 - 6 980 74
AK397667.1 - 6 - 6 936 74
AK399356.1 - 6 - 6 933 74
AK400084.1 - 6 - 6 955 74
AY864612.1 - 2 - 2 171 74
AY997700.1 - 6 - 6 754 74

View Full Product Details