Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
WAP
Entrez Gene ID
396835
Gene Name
whey acidic protein
Gene Aliases
-
Chromosome Location
Chr.18: 50359599 - 50361906 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213841.2 NP_999006.1 3 - 3 251 124
XM_013985798.2 XP_013841252.1 - 538 124
GenBank mRNA AF034646.1 - 3 - 3 205 124
AJ000221.1 - 3 - 3 251 124

View Full Product Details