Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDK5R1
Entrez Gene ID
100113467
Gene Name
cyclin dependent kinase 5 regulatory subunit 1
Gene Aliases
-
Chromosome Location
Chr.12: 42456063 - 42494494 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101816.2 NP_001095286.2 1 - 1 851 101
XM_021066022.1 XP_020921681.1 - 932 101
XM_021066023.1 XP_020921682.1 - 932 101
GenBank mRNA EU041702.2 - 1 - 1 851 101

View Full Product Details