Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDCD1LG2
Entrez Gene ID
574057
Gene Name
programmed cell death 1 ligand 2
Gene Aliases
PDL2
Chromosome Location
Chr.1: 216564428 - 216600333 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025220.1 NP_001020391.1 3 - 4 626 91
GenBank mRNA AK230902.1 - 3 - 4 668 91
AK344340.1 - 2 - 3 632 91
AY837781.1 - 3 - 4 626 91

View Full Product Details