Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTLA4
Entrez Gene ID
397286
Gene Name
cytotoxic T-lymphocyte associated protein 4
Gene Aliases
-
Chromosome Location
Chr.15: 107283081 - 107288041 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214149.1 NP_999314.1 2 - 3 476 82
GenBank mRNA AF220248.1 - 2 - 3 476 82
AF281633.1 - 2 - 3 463 82

View Full Product Details