Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IRF6
Entrez Gene ID
397521
Gene Name
interferon regulatory factor 6
Gene Aliases
-
Chromosome Location
Chr.9: 133164360 - 133184045 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214278.1 NP_999443.1 2 - 3 374 123
XM_005667890.3 XP_005667947.1 - 668 123
XM_005667891.3 XP_005667948.1 - 729 123
XM_005667892.3 XP_005667949.1 - 739 123
XM_005667893.3 XP_005667950.1 - 568 123
XM_013979999.2 XP_013835453.1 - 3633 123
XM_021102240.1 XP_020957899.1 - 537 123
GenBank mRNA AF327368.1 - 2 - 3 374 123

View Full Product Details