Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSHB
Entrez Gene ID
397658
Gene Name
thyroid stimulating hormone subunit beta
Gene Aliases
TSH-BETA
Chromosome Location
Chr.4: 105553070 - 105558769 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214368.2 NP_999533.1 1 - 2 166 90
XM_005663554.3 XP_005663611.1 - 267 90
XM_021088723.1 XP_020944382.1 - 185 90
GenBank mRNA U39816.1 - 1 - 2 169 90

View Full Product Details