Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GZMA
Entrez Gene ID
100526762
Gene Name
granzyme A (granzyme 1, cytotoxic T-lymphocyte-associated serine esterase 3)
Gene Aliases
-
Chromosome Location
Chr.16: 34282160 - 34290422 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198926.1 NP_001185855.1 1 - 2 88 79
GenBank mRNA HQ225831.1 - 1 - 2 88 79

View Full Product Details