Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OIT3
Entrez Gene ID
100513952
Gene Name
oncoprotein induced transcript 3
Gene Aliases
-
Chromosome Location
Chr.14: 75726353 - 75753461 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244899.1 NP_001231828.1 7 - 8 1471 90
XM_021072055.1 XP_020927714.1 - 1489 90
GenBank mRNA AK346287.1 - 7 - 8 1471 90

View Full Product Details