Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRMT10A
Entrez Gene ID
100512243
Gene Name
tRNA methyltransferase 10A
Gene Aliases
RG9MTD2
Chromosome Location
Chr.8: 120879067 - 120917442 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244146.2 NP_001231075.1 8 - 9 1072 69
XM_005653601.2 XP_005653658.1 - 1927 69
XM_005653603.3 XP_005653660.1 - 1338 69
XM_005653605.2 XP_005653662.1 - 1805 69
XM_005653610.3 XP_005653667.1 - 1554 69
XM_013998341.1 XP_013853795.1 - 1703 69
XM_021100333.1 XP_020955992.1 - 2843 69
XM_021100334.1 XP_020955993.1 - 1737 69
XM_021100335.1 XP_020955994.1 - 2015 69
XM_021100336.1 XP_020955995.1 - 1562 69
XM_021100337.1 XP_020955996.1 - 1569 69
XM_021100338.1 XP_020955997.1 - 1663 69
XM_021100339.1 XP_020955998.1 - 1513 69
XM_021100340.1 XP_020955999.1 - 1477 69
XM_021100341.1 XP_020956000.1 - 1395 69
XM_021100342.1 XP_020956001.1 - 1206 69
GenBank mRNA AK238426.1 - 8 - 9 1073 69

View Full Product Details