Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHCHD3
Entrez Gene ID
414383
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 3
Gene Aliases
-
Chromosome Location
Chr.18: 16052611 - 16348998 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244176.1 NP_001231105.1 7 - 8 818 102
XM_021078529.1 XP_020934188.1 - 839 102
GenBank mRNA AK231938.1 - 7 - 8 803 102
AK234688.1 - 7 - 8 755 102
AK348653.1 - 7 - 8 757 102
AK393133.1 - 7 - 8 806 102
AY610152.1 - 7 - 8 818 102

View Full Product Details