Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LIN7B
Entrez Gene ID
100512352
Gene Name
lin-7 homolog B, crumbs cell polarity complex component
Gene Aliases
-
Chromosome Location
Chr.6: 54309552 - 54313138 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244232.1 NP_001231161.1 3 - 4 266 99
GenBank mRNA BF190116.1 - 3 - 4 246 99
DY423750.1 - 3 - 4 459 99
EW092492.2 - 2 - 3 139 99

View Full Product Details