Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MIR1
- Entrez Gene ID
-
100316581
- Gene Name
- microRNA mir-1
- Gene Aliases
-
MIR1A, ssc-mir-1, ssc-mir-1a
- Chromosome Location
-
Chr.17: 61894036 - 61894143 on Build Sscrofa11.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_035375.1 |
|
1 - 1 |
54 |
101 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
translation initiation factor IF-2-like |
LOC110257484 |
|
-2.2kb
|
Anti-sense |
Chr. - 61885480 - 61891849 on Build Sscrofa11.1 |
Downstream |
microRNA mir-133a-1 |
MIR133A-1 |
MIR133A, ssc-mir-133a, ssc-mir-133a-1 |
10.6kb
|
Sense |
Chr. - 61904781 - 61904883 on Build Sscrofa11.1 |
View Full Product Details