Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
stoml2
Entrez Gene ID
448051
Gene Name
stomatin like 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016683457: 226175 - 229106 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004808.1 NP_001004808.1 - 785 70
GenBank mRNA BC074573.1 - - 785 70
CR761170.2 - - 777 70

View Full Product Details