Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
psmd8
Entrez Gene ID
448645
Gene Name
proteasome 26S subunit, non-ATPase 8
Gene Aliases
hip6, hypf, nin1p, psmd8-a, psmd8-b, rpn12
Chromosome Location
Chr.8: 58968336 - 58976735 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005073.1 NP_001005073.1 4 - 5 481 106
XM_012968255.2 XP_012823709.1 4 - 5 598 106
GenBank mRNA BC076990.1 - 4 - 5 481 106
CU075836.1 - 4 - 5 468 106

View Full Product Details