Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
bcap31
Entrez Gene ID
394579
Gene Name
B cell receptor associated protein 31
Gene Aliases
6c6-ag, bap31, cdm, dxs1357e
Chromosome Location
Chr.8: 3214079 - 3228395 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203651.1 NP_988982.1 3 - 4 294 68
GenBank mRNA BC061339.1 - 3 - 4 294 68

View Full Product Details