Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- bzw1
- Entrez Gene ID
-
394895
- Gene Name
- basic leucine zipper and W2 domains 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 49243707 - 49253282 on Build UCB_Xtro_10.0
- Gene Symbol
- tent4b
- Entrez Gene ID
-
100491336
- Gene Name
- terminal nucleotidyltransferase 4B
- Gene Aliases
-
papd5
- Chromosome Location
-
Chr.4: 46509637 - 46564827 on Build UCB_Xtro_10.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC168133.1 |
- |
- |
2076 |
103 |
View Full Product Details