Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ythdf1
Entrez Gene ID
395029
Gene Name
YTH N(6)-methyladenosine RNA binding protein 1
Gene Aliases
daca-1
Chromosome Location
Chr.Un|NW_016684343: 1912 - 5557 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204061.1 NP_989392.1 1 - 2 325 76
GenBank mRNA BC064856.1 - 1 - 2 325 76

View Full Product Details