Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cidec
Entrez Gene ID
493464
Gene Name
cell death inducing DFFA like effector c
Gene Aliases
cide-3, fsp27
Chromosome Location
Chr.4: 127654100 - 127662709 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008101.1 NP_001008102.1 5 - 6 525 81
XM_012960954.2 XP_012816408.1 4 - 5 1069 81
XM_012960955.2 XP_012816409.1 6 - 7 824 81
XM_012960956.2 XP_012816410.1 4 - 5 513 81
XM_012960957.2 XP_012816411.1 4 - 5 498 81
XM_018093023.1 XP_017948512.1 5 - 6 722 81
GenBank mRNA BC081304.1 - 5 - 6 525 81
CR761715.2 - 5 - 6 533 81

View Full Product Details