Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
irx2
Entrez Gene ID
493475
Gene Name
iroquois homeobox 2
Gene Aliases
Xiro2, iro2
Chromosome Location
Chr.6: 36741628 - 36746801 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008112.1 NP_001008113.1 4 - 5 1836 86
GenBank mRNA BC081317.1 - 4 - 5 1836 86

View Full Product Details