Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
btf3l4
Entrez Gene ID
496686
Gene Name
basic transcription factor 3-like 4
Gene Aliases
-
Chromosome Location
Chr.4: 86385028 - 86398679 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011243.1 NP_001011243.1 5 - 6 482 102
XM_012960996.2 XP_012816450.1 5 - 6 528 102
XM_012960997.2 XP_012816451.1 5 - 6 527 102
XM_012960998.2 XP_012816452.1 5 - 6 527 102
XM_012960999.2 XP_012816453.1 5 - 6 460 102
GenBank mRNA BC087817.1 - 5 - 6 482 102
CR848347.2 - 4 - 5 436 102

View Full Product Details