Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
twist1
Entrez Gene ID
395054
Gene Name
twist family bHLH transcription factor 1
Gene Aliases
X-twi, Xtwi, Xtwist, acs3, bpes2, bpes3, scs, twist, twist1-a, twist1-b
Chromosome Location
Chr.6: 43550969 - 43553328 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204084.2 NP_989415.1 1 - 2 670 78
GenBank mRNA BC074558.1 - 1 - 2 670 78
CR760828.2 - 1 - 2 665 78

View Full Product Details