Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
haus2
Entrez Gene ID
548581
Gene Name
HAUS augmin like complex subunit 2
Gene Aliases
cep27
Chromosome Location
Chr.8: 85295509 - 85304066 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015864.1 NP_001015864.1 1 - 2 116 49
GenBank mRNA BC090378.1 - 1 - 2 116 49

View Full Product Details