Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
figla
Entrez Gene ID
549096
Gene Name
folliculogenesis specific bHLH transcription factor
Gene Aliases
bhlhc8, figalpha, pof6
Chromosome Location
Chr.3: 112326788 - 112337655 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016342.2 NP_001016342.1 6 - 7 716 104
XM_012959005.2 XP_012814459.1 6 - 7 825 104
GenBank mRNA BC155532.1 - 5 - 6 689 104
CR761381.2 - 6 - 7 716 104

View Full Product Details