Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sae1
Entrez Gene ID
549624
Gene Name
SUMO1 activating enzyme subunit 1
Gene Aliases
Aos, Aos1p, Sua1p, aos1, hspc140, sua1
Chromosome Location
Chr.8: 55889927 - 55915445 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016870.2 NP_001016870.1 2 - 3 204 68
GenBank mRNA BC135749.1 - 2 - 3 249 68
CR848627.2 - 2 - 3 204 68

View Full Product Details