Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
prelid2
Entrez Gene ID
549676
Gene Name
PRELI domain containing 2
Gene Aliases
-
Chromosome Location
Chr.3: 30146972 - 30192382 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016922.2 NP_001016922.1 4 - 5 401 94
GenBank mRNA BC091072.1 - 4 - 5 401 94
CR848108.2 - 4 - 5 433 94

View Full Product Details