Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
b3gat3
Entrez Gene ID
594895
Gene Name
beta-1,3-glucuronyltransferase 3
Gene Aliases
glcati
Chromosome Location
Chr.4: 41607514 - 41618366 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030349.1 NP_001025520.1 4 - 5 814 96
XM_012954327.2 XP_012809781.1 6 - 7 1198 96
XM_012954328.2 XP_012809782.1 5 - 6 1016 96
XM_012954329.2 XP_012809783.1 6 - 7 1186 96
XM_012954330.2 XP_012809784.1 5 - 6 1004 96
XM_012954331.2 XP_012809785.1 5 - 6 996 96
GenBank mRNA AJ890837.1 - 4 - 5 814 96
BC121200.1 - 6 - 7 1101 96

View Full Product Details